Specialty Cleaning Tools and Wipes for Efficient and Ergonomic Sanitization
Porcess cabinents, curved surfaces, and spaces between, under, and behind equipment need special cleaning attention. Micronova's range of specialty cleaning tools and wipes simplify the task. From biosafety cabinets to lyophilizers, round vessels to curved joints, and plastic strip curtains to soft walls, there are options for removing contaminants for the most challenging of conditions.
MegaSwat™ Mops Blade Style Mop
Single ply and foam lined blade style mops easily glide under and behind equipment. Adapters and mop covers are available in a range of widths, lengths and materials.
Econo CurtainCleaner™ Tool and Covers
Cleans both sides of strip curtain at same time allowing for a more efficient and effective cleaning.
Isolator Tool™ and Covers
Ergonomic and effective cleaning for isolators, hoods, RABS and other small cabinets. 360 swivel and range of fabrics for cleaning or disinfection.
LyoTool™ Dual Plate Mop
Cleans top and bottom of shelves simultaneously. Adjustable spring loaded shelves provide consistent pressure and release of cleaning agent.
PolyMitt™ Cleaning Mitt Wipes
Durable cleaning mitt can be used wet or dry. Design allows for better surface coverage than a standard wipe.
Tank Tool™ for Exterior and Interior Tank Contours
Flexible adapter conforms to tank with pressure against surface for good cleaning contact. Easily cleans tank and cylinder sides, bottoms and interiors; exterior of vessels.