Stevanato Group EZ-fill® Vials from DWK Life Sciences
Ready-to-Use Sterile Vials designed to accelerate the drug development process and to increase operational flexibility in Aseptic Filling.
Ideal for drug development process, from first line trials through to commercialization, as well as small batch manufacturing.
Remove cost, time and complexity associated with particulate cleaning, depyrogenation and sterilization through the use of Ready-to-Use (RTU) vial solution.
Improve product quality and patient safety with RTU vials packed in secondary packaging that eliminates glass-to glass contact.
Reduced time to market through use of world’s most widely adopted RTU vial platform, ensuring quick scalability from early stage development to commercial production.
About Stevanato Group EZ-fill® Vials
- Particulate clean, depyrogenated and sterile vials in a Nest and Tub or Partitioned Trays
- ISO 8362-1 2R, 4R, 6R, 10R, and 20R vials, manufactured from borosilicate type 1, 51 expansion glass, with European Blowback
- Secondary packaging is designed for direct entry into aseptic filling operations and to prevents glass-to-glass contact for higher patient safety and product quality
- Nested configurations designed for direct entry into flex fill, robotic, and combi filling operations
- Tray configurations suitable for small batch and in-line filling operations
- World’s most widely adopted RTU platform ensures compatibility with different fill & finish processes, enabling the adoption of multi-product lines
Why Stevanato Group EZ-fill® Vials?
Ready-to-Use primary packaging helps to accelerate the drug development journey by increasing operational flexibility in Aseptic Filling and enabling a move towards personalized medicines and biologics.
Stevanato Group’s EZ-fill® Vials help streamline operations by removing steps such as cleaning, depyrogenation and sterilization so you can concentrate on your core competencies of drug development and production.
At lab scale, companies can rely on Stevanato Group EZ-fill® vials to increase reliability in the development process by enabling access to commercial scale quality at development stage cost.
Stevanato Group’s EZ-fill® Vials are also the world’s most widely adopted Ready-to-Use vial standard and their adoption at the early stages of drug development will reduce the time, cost and complexities of the future transition to commercial scale production.
Webinar
From pipeline to patient: Find out how Ready-to-Use vials can accelerate the drug development process.
Learn more