Skin and Glove Cleansing and Sanitization
Micronova's hand hygiene products are an important part of keeping employees and products clean and safe from debris, microbials, and viruses. Products range from cleansing wipes to hand soaps and glove sanitizers which can be used with VWR Touch Free Dispenser.
NovaClenz™ Instant Hand and Glove Sanitizer
Highly effective, fast-acting instant hand sanitizer ideal for use under gloves. Contains 65% minimum ethanol which helps to prevent viral transfer. Requires no sink, water, or towels. Effective against bacteria, including: pseudonomas, salmonella, and staphylococcus and reducing nosocomial infections.