Specialty Brushes for Labware and Process Equipment
Brushes are essential for cleaning small diameter items, process equipment, portable tanks, and storage vessels. With a robust range of styles and sizes, includng flexible stainless steel or polypropylene cores, and diameters, from 1/4 inch to 5 inches, Micronova brushes meet the lab and production challenges.
Bottle Brush
Color-coded stiff polyester bristles brushes with polypropylene handles for process equipment and parts. BPA-free base resin, FDA approved materials of construction.
Cylinder Brush
Dome shaped, lightweight yet durable polyester bristle brush. Attaches to threaded handles for extended reach. BPA-free base resin, FDA approved materials of construction.
Kettle Brush
Oblong shaped, lightweight, and durable brush. Attaches to threaded handles. Polyester bristles, BPA-free base resin, FDA approved materials of construction.
Valve Brush
Large diameter, durable polyester bristle brush with attached polypropylene handle. Excellent for bowl and small vessels. BPA-free base resin, FDA approved materials of construction.
FlexBrush™ Curved Brush
A hook-shaped nylon bristle brush with stainless steel core. Its flexible core allows user to shape brush to custom applications including overhead piping and equipment interiors. Attaches to threaded handles.
Plastic Tube Brush
100% polypropylene tube brushes. Ideal for cleaning finely polished stainless steel parts or tubing without scratching. A flexible core that glides through angled piping and fittings.
Stainless Steel Wire Brushes
Durable stainless steel wire brushes with nylon bristles. Available in diameters of 1/4 to 2 inches and lengths from 12 to 72 inches.
Drain Brush
A stiff black bristle, replaceable brush head, and a threaded plastic handle specifically designed and labeled for cleaning floor drains.