NovaClean™ Family of Cleanroom Detergents
Non-ionic detergents remove particles and residues common in cleanrooms and critical laboratories. With low salts and metals, in the ppb range, these cleanroom detergents are ideal for semiconductors, electronics, medical device, and pharmaceutical industries.
NovaClean™ Lab & Glass Cleanroom Detergent
A non-solvent based detergent formulated to be applied directly to cleanroom work surfaces including glass, stainless steel, epoxy, acrylic, and painted surfaces. Free-rinsing and biodegradable.
NovaHol™ Cleanroom Cleaner
NovaHol™ with the same pure formula as original NovaClean™ detergent, has added isopropyl alcohol for a faster dry time. NSF rated, category A1 (general cleaners).