Isolator cleaning tool: Mop hardware, replacement head and handle
NEW!
Specially engineered to work with isolators and other small, hard to reach areas
- Easy to manoeuvre, lightweight handles offer one-handed operation
- Low profile, swivel head allows you to get into hard to reach places
- Engineered for quick cover changes during operation, the padded head allows for consistent mopping across different surfaces without losing cleaning power
- The rounded edges clean isolator corners and prevent scratches or damage to surfaces
- Available with different types of mop handles and mop covers, sterile and non sterile
Log in to see your contract pricing and availability.