![](/m-ni.vwr.com/en_NI/images/contec_logo.jpg)
Consumable products for endotoxin control
![](/m-ni.vwr.com/en_NI/images/contec_endotoxin_control_main.gif)
VWR solutions to reduce bioburden and control endotoxin levels in your manufacturing process
- Sterile alcohols and disinfectants (availability may vary per country so contact your local VWR customer service for more information)
- Amplitude™ Kappa™ Sterile LE, dry non woven wipes, cost effective, double bagged in packs of 25 pieces, with an endotoxin limit of 20 EU per wipe
- Polynit Heatseal Sterile LE, dry polyester wipes, with sealed edges, half folded and triple bagged in packs of 10 pieces, with an endotoxin limit of 1 EU per wipe
- PROSAT® Sterile PS-911 LE, polypropylene wipes pre-saturated with 70% pharma grade IPA and 30% deionised water, double bagged in reclosable pouches of 30 pieces, with an endotoxin limit of 20 EU per wipe
- PROSAT® Sterile Polynit Heatseal LE, sealed edge polyester wipes pre-saturated with 70% pharma-grade IPA and 30% Water for Injection (USP), triple bagged in reclosable pouches of 10 pieces, with an endotoxin limit of 1 EU per wipe
Log in to see your contract pricing and availability.
Description | Material | Dry/pre-saturated | Packed | Pk | Cat. No. | Price | Quantity | |
---|---|---|---|---|---|---|---|---|
Amplitude™ Kappa™ Sterile LE wipes; 23x23 cm, <20 EU/wipe | Non woven polyester/lyocell blend | Dry | Double bagged, 40x25 wipes | 1000 | 115-1863E | |||
Amplitude™ Kappa™ Sterile LE wipes; 30x30 cm, <20 EU/wipe | Non woven polyester/lyocell blend | Dry | Double bagged, 40x25 wipes | 1000 | 115-1864E | |||
Polynit Heatseal Sterile LE wipes; 23x23 cm, <1 EU/wipe | Knitted polyester with sealed edges | Dry | Triple bagged, 50x10 wipes | 500 | 115-2438E | |||
PROSAT® Sterile PS-911 LE wipes; 23x28 cm, <20 EU/wipe | Non woven polypropylene | 70% pharma grade IPA 30% deionised water | Double bagged, 36x30 wipes | 1080 | 115-2439 | |||
PROSAT® Sterile Polynit Heatseal LE wipes; 23x23 cm, <1 EU/wipe | Knitted polyester with sealed edges | 70% pharma grade IPA 30% water for injection | Triple bagged, 55x10 wipes | 550 | 115-1424E | |||
PROSAT® Sterile Polynit Heatseal LE wipes; 30x30 cm, <1 EU/wipe | Knitted polyester with sealed edges | 70% pharma grade IPA 30% water for injection | Triple bagged, 30x10 wipes | 300 | 115-2440 |