Sterilization monitoring
3M is one of the world leaders in sterilization monitoring solutions. Our complete portfolio of differentiated, clinically preferred products can help you streamline your operations and standardise your protocols.
Products like 3M™ Attest™ Super Rapid Readout Biological Indicators can significantly reduce the time required to see critical information about the sterilization process. Our Type 5 3M™ Comply™ (SteriGage™) Chemical Integrator features moving front ink technology, with an easy to interpret ‘Accept or Reject’ result and 3M™ Comply™ Lead Free Steam Indicator Tape features 3M’s renowned adhesive technologies to ensure pack security, while providing the ‘at a glance’ information you need to quickly confirm sterilisation exposure.
Featured Product
3M™ Electronic Test System For Steam Sterilisers (ETS)™
ETS is an easy to use electronic test device which provides comprehensive, objective, independent information about the critical variables of the steam sterilisation process in which it is used. ETS provides an innovative alternative to the traditional Bowie and Dick test pack and its derivatives.
3M™ Attest™ Super Rapid Readout Biological Indicator 1492V For Steam Sterilisation
The Biological Indicator provides BI results in 24 minutes in 3M Attest 490 Auto-reader. It is appropriate for dynamic air removal (pre-vacuum and SFPP): 135˚C (275˚F) 3 min. It meets ISO performance requirements for biological indicators and contains viable organisms that provide a direct measure of lethality.
3M™ Attest™ Auto-reader, 490
This dual-Auto-reader designed for use with 3M™ Attest™ Biological Indicators 1491, 1492V and 1295. It has Intelligent display with ‘time to result’ countdown feature. The display is viewable remotely using your PC. Use the same 3M™ Web Application you use now for real time BI status and results for digital record keeping.
3M™ Comply™ SteriGage™ Steam Chemical Integrating Indicators
3M Comply SteriGage Steam Chemical Integrators meet the performance requirements of EN ISO 11140- 1. 3M Comply SteriGage Steam Chemical Integrators are designed to accurately detect small changes in all defined critical sterilisation parameters across the entire common sterilisation range in gravity and pre-vac steam sterilisation processes.
3M™ Comply™ Steam Indicator Tape
A reliable steam indicator tape that provides immediate information if the pack has been exposed to adequate steam sterilisation. The chemical indicator lines will turn dark when exposed to a steam sterilisation process.