Fall protection
3M offers everything you need for working at heights. Discover an extensive range of reliable, comfortable, innovative and high quality fall arrest equipment and solutions; anchorages, harnesses, connectors, rescue and evacuation equipment, temporary and permanent vertical and horizontal lifeline systems, confined space equipment, professional training services and more.
See our webinar about fall protectionOur mission is to provide you with a comprehensive solution. That’s why we have a team of specialist application engineers who can advise on risk assessment, project development and the most suitable working at height solution for your needs.
At 3M we combine technical innovation and expertise with a passion for manufacturing the highest quality products. 3M™ Fall Protection elevates worker safety to new heights with the proven and trusted DBI-SALA® and Protecta® brands of fall protection equipment and solutions. These solutions include innovative harnesses, lanyards, anchors, horizontal lifelines, access systems, rescue and evacuation equipment, confined space solutions, all PPE or personal protective equipment needed for personal fall arrest protection and much more!
We meet the world's most rigorous standards and codes including CE and ANSI, giving you maximum freedom and confidence. The sky’s the limit!
Featured Product
DBI-SALA® Short Reach Davit System
3M™ DBI-SALA® Confined Space HC (High Capacity) Short Reach Davit System is constructed of powder-coated aluminium, and features a modular design for easy transport and storage. Our system is adjustable, providing flexibility during installation. This product is certified to EN795:2012, CEN/TS 16415:2013, EN1496:2012, and meets the requirements of machinery directive 2006/42EC for material handling.
Resources
DBI-SALA® Delta Comfort Rescue Harness
3M™ DBI-SALA® Delta™ Comfort Rescue Harness is one of our most popular harnesses featuring comfortable padding, with an ergonomic fit and feel, lightweight buckles and high strength webbing. It has a no-tangle triangular pattern, which creates a frame that allows the harness to be easily donned.
DBI-SALA® Sealed-Blok™ SRL with Retrieval Winch
Our 3M™ DBI-SALA® Sealed-Blok™ Retrieval Self-Retracting Lifeline, has been designed to stand up to the harshest environments. With all the features of our standard Sealed-Blok™ plus a 3-way retrieval winch. The heavy-duty design is corrosion resistant, with stainless steel end-plates and aluminium housing. For reliability and quality fall protection in applications such as oil and gas rigging, confined space, saltwater environments, dirty environments and sewage plants, Sealed-Blok™ has many years of service in the field.
D - 3M™ Protecta® Tripod
When it comes to confined space entry/rescue, it is essential that equipment operates perfectly and quickly. Lightweight, portable Protecta® aluminium tripods are ideal for confined space manhole entry and retrieval. They’re easily transported from one location to another and can be set up by one worker. Attach one or two mechanical devices such as a winch or 3 way self retracting lifeline and you’re ready to go. Our tripods are loaded with features such as locking legs, simple detent pin style adjustment, quick mount mechanical device bracket and skid resistant tread plate soles on feet.