Universal Fit Pipette Tips
Thermo Scientific™ ART™ pipette tips provide a complete selection of barrier, non-filtered and specialty tips. It has never been easier to find a tip to fit virtually any pipette or application in your laboratory. All deliver proven ART quality, performance and reliability, backed by the highest Thermo Scientific standards and worldwide support.
From 10 μL to 10 mL barrier and non-filtered tips, to gel loading and wide bore specialty products – every tip is certified to meet the industry's highest standards for molding precision and clarity, and are certified free of RNase, DNase, ATP, and Pyrogens. And each meets the exact specifications of its respective pipette brand manufacturer.
Challenge our Barrier Tips at no cost
Thermo Scientific ART™ Barrier tips
The Thermo Scientific™ ART™ self-sealing barrier provides 100% protection against aerosols and liquid contamination. A complete product offering including racked and reload packaging options as well as gel loading, extended length, solvent safe and wide pore specialty tips make it easy to find a tip to fit virtually any pipette or application in the laboratory. Using advanced low-retention technology, the hydrophobic inner surface of ART™ Low retention tips significantly reduces sample loss to adhesion and maximizes sample recovery.
Thermo Scientific ART™ Non-filtered tips
Complete selection of non-filtered and specialty tips deliver proven ART™ quality, performance and reliability. ART™ Non-filtered tips are ideal choice to any pipette or application in the laboratory. Available as gel loading, extended length, wide pore and low retention specialty tips as well as racked, reload and bulk packaging options.