Manual Pipetting Systems
Introducing the new Finnpipette F1 and F1-ClipTip pipettes
The evolution of excellence in pipetting takes a huge step forward – thanks to your valuable feedback. Each suggestion turned into a key feature that makes your life in the lab safer.
- Silent, effortless volume adjustment with over 50% lower forces compared to earlier models
- A textured volume adjustment knob and optional grip strip improve grip
- Lowered pipetting forces now reduce Repetitive Stress Injury (RSI) even further without compromising results.
Upgrade to exceptional performance with comfort, redefined.
ClipTip Manual Pipetting Systems
Thermo Scientific™ F1-ClipTip™ pipettes transform daily pipetting with innovative Thermo Scientific ClipTip interlocking technology, which ensures a complete seal on every channel with minimal tip attachment and ejection force. Once attached, the tips are locked firmly in place and will not loosen or fall off regardless of application pressure.
Finnpipette Manual Pipetting Systems
Thermo Scientific Finnpipette™ F1 pipettes offers superior ergonomics, and performance with a lightweight design combined with low tip attachment and ejection forces. Finnpipette F2 pipette models are fully autoclavable, and high-quality materials offer durability even in the most demanding applications. Choose from a full range of Finntip™ Flex or Finntip pipette tips for ideal performance.
![]() |
![]() |
Literature
Request literature or download PDFs from Thermo Scientific...
Promotions
View a comprehensive list of the latest Thermo Scientific promotions...