BOD Photometric Method
The BOD test is a photometric cell test from Merck
- Calibrated and standard compliant analysis reagents for security
- Bar code system enables rapid and simple operation
- Comprehensive documentation of the quality control for every test
- 5 days incubation still required but the analysis has fewer handling steps than other methods