Dilution BOD
BOD measurement official method according to EN 1899-1 and DI EN 1899-2, official EPA method.
- Simple sample management
- Automatic BOD calculation
- EPA approved
Self-stirring oxygen sensor StirrOx® G for oxygen determination in Karlsruhe bottles allows:
- Constant flow for high reproductibility
- No zero point calibration necessary
- Immediately ready for measuring