Anti-6A3-IgG4 Recombinant Antibody

Supplier: ProSci Inc.
10-443
PRSI10-443EA 610 GBP
PRSI10-443
Anti-6A3-IgG4 Recombinant Antibody
Antibodies
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2-G4-kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR