Microcystin-LA
Supplier: ENZO LIFE SCIENCES
ALX350096C100 ALX350096C025 ALX350096C025 ALX350096C100
ALEXALX350096C100EA 473 GBP
ALEXALX350096C100 ALEXALX350096C025 ENZOALX350096C025 ENZOALX350096C100
Microcystin-LA
Proteins and Peptides
Inhibitor of PP2A and calcineurin
Analog of microcystin-LR (Prod. No. ALX-350-012) with Ala substituted in place of Arg. Inhibits protein phosphatase 2A (PP2A) and protein phosphatase 3 (PP3) more potently than protein phosphatase 1 (PP1). Ozonation of toxic water samples does lead to a complete loss of the microcystins and their toxicity.
Isolated from Microcystis aeruginosa.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef9d3082ea6c92cabbac44b1d9af720c26cd57b6&vid=a5a5ac66ef6547fc41e0c401711ab63ee23f742a&prid=web&clid=SAPDAM