Anti-CRYAA Rabbit Polyclonal Antibody

Supplier: ENZO LIFE SCIENCES
ADI-SPA-221-F ADI-SPA-221-D ADISPA221D ADISPA221F
ASSAADI-SPA-221-FEA 434 GBP
ASSAADI-SPA-221-F ASSAADI-SPA-221-D ENZOADISPA221D ENZOADISPA221F
Anti-CRYAA Rabbit Polyclonal Antibody
Antibodies
Alpha-crystallins, which are part of the small Heat shock family members, are major water-soluble proteins present in the lens of the mammalian eye. Phosphorylation of serine residues which occurs during development and in response to stress, is intimately linked with its function. Chaperone activity requires, and is modulated by, oligomerization and is limited to binding unfolded intermediates to prevent irreversible aggregation.

Type: Primary
Antigen: CRYAA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR