Anti-HDJ2 Rabbit Polyclonal Antibody

Supplier: ENZO LIFE SCIENCES
ADISPA405050 ADISPA405200
ENZOADISPA405050EA 193 GBP
ENZOADISPA405050 ENZOADISPA405200
Anti-HDJ2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HDJ2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR