Anti-E6AP Mouse Monoclonal Antibody

Supplier: ENZO LIFE SCIENCES
BMLPW05350025 BMLPW05350100
ENZOBMLPW05350025EA 99.2 GBP
ENZOBMLPW05350025 ENZOBMLPW05350100
Anti-E6AP Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Western Blot

Recognizes human E6AP.

Type: Primary
Antigen: E6AP
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR