Anti-PBA1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ENZOBMLPW04300025EA 171 GBP
ENZOBMLPW04300025 ALEXBMLPW04300025 ENZOBMLPW04300100 ALEXBMLPW04300100
Anti-PBA1 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Western Blot

Recognizes Arabidopsis proteasome 20S (PBA1/beta1 subunit).

Type: Primary
Antigen: PBA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Arabidopsis thaliana
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR