Anti-STX6 Mouse Monoclonal Antibody

Supplier: ENZO LIFE SCIENCES
ADIVAMSV025D ADIVAMSV025F
ENZOADIVAMSV025DEA 263 GBP
ENZOADIVAMSV025D ENZOADIVAMSV025F
Anti-STX6 Mouse Monoclonal Antibody
Antibodies
Syntaxins are type II integral membrane proteins localized to membranes that regulate vesicle transport, protein maturation, and exocytosis. Syntaxin 1A resides in the nerve terminals of sensory neurons and nerve fibers reaching small blood vessels. Syntaxin 2 localizes to the apical plasma membrane and regulates epithelial-mesenchymal interactions. Syntaxin 6 is primarily localized to the trans-Golgi network, and partially co-localizes with AP-1 on clathrin-coated membranes. Syntaxin 13 is ubiquitously expressed and localizes to the tubular early and recycling endosomes, as well as endosomal vacuoles.

  • For Immunoprecipitation, Western Blot

Recognizes human, mouse, rat and hamster Syntaxin 6. Detects a band of ~35kDa by Western blot.

Type: Primary
Antigen: STX6
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR