Anti-GS28 Mouse Monoclonal Antibody

Supplier: ENZO LIFE SCIENCES
ADIVAMPT047D ADIVAMPT047F
ENZOADIVAMPT047DEA 353 GBP
ENZOADIVAMPT047D ENZOADIVAMPT047F
Anti-GS28 Mouse Monoclonal Antibody
Antibodies
Golgi SNARE of 28 kDa (GS28), also known as p28 or GOS28, is a 28 kDa integral membrane protein on the surface of the Golgi apparatus that serves as a t-SNARE in ER to Golgi transport. The amino-terminal of GS28 is exposed to the cytosol and is anchored to the cis Golgi via a 20 amino acid carboxyl-terminal hydrophobic tail. GS28 co-immunoprecipitates complexes consisting of Syntaxin 5, Rbet1, Membrin, Rsec22, and Rsly1, and is therefore implicated in ER to-Golgi or intra-Golgi vesicle transport.

  • For Western Blot, EM

Recognizes mouse, rat, bovine, dog, chicken, hamster, pig, rabbit and sheep GS28. Detects a band of ~28kDa by Western blot.

Type: Primary
Antigen: GS28
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR