Anti-LAMP1 Mouse Monoclonal Antibody

Supplier: ENZO LIFE SCIENCES
ADIVAMEN001D ADIVAMEN001F
ENZOADIVAMEN001DEA 264 GBP
ENZOADIVAMEN001D ENZOADIVAMEN001F
Anti-LAMP1 Mouse Monoclonal Antibody
Antibodies
Lysosome-associated membrane proteins (LAMP1 and LAMP2) are type I integral membrane protein that are transported from the trans-Golgi network to endosomes and then lysosomes. Following activation of platelets, T-cells, neutrophils, or endothelium, LAMP1 and LAMP2 are expressed on the cell surface. Cell surface LAMP1 and LAMP2 have been shown to promote adhesion of human peripheral blood mononuclear cells (PBMC) to vascular endothelium, and loss of LAMP2 expression is associated with impaired autophagy observed in Danons disease.

  • For Immunocytochemistry, Immunofluorescence, Immunoprecipitation, EM

Recognizes human, rat and hamster LAMP1. Detects a band of ~120kDa by Western blot.

Type: Primary
Antigen: LAMP1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR