Anti-PDI Mouse Monoclonal Antibody

Supplier: ENZO LIFE SCIENCES
ADI-SPA-891-F ADI-SPA-891-J ADISPA891D ADISPA891F ADISPA891J
ASSAADI-SPA-891-FEA 499 GBP
ASSAADI-SPA-891-F ASSAADI-SPA-891-J ENZOADISPA891D ENZOADISPA891F ENZOADISPA891J
Anti-PDI Mouse Monoclonal Antibody
Antibodies
The mammalian PDI (Protein disulfide-isomerase) family encompasses several highly divergent proteins which are involved in the processing and maturation of secretory proteins in the endoplasmic reticulum by catalyzing the rearrangement of disulfide bonds.

  • For Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot, EM

Recognizes human, mouse, rat, bovine, dog, chicken, guinea pig, hamster, monkey, pig, rabbit, sheep and Xenopus PDI. Detects a band of ~58kDa by Western blot.

Type: Primary
Antigen: PDI
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR