Anti-HSP60 Mouse Monoclonal Antibody

Supplier: ENZO LIFE SCIENCES
ADI-SPA-829-F ADI-SPA-829-D ADISPA829D ADISPA829F
ASSAADI-SPA-829-FEA 486 GBP
ASSAADI-SPA-829-F ASSAADI-SPA-829-D ENZOADISPA829D ENZOADISPA829F
Anti-HSP60 Mouse Monoclonal Antibody
Antibodies
Hsp60 is a member of the chaperonin family of heat shock proteins, with homologs functioning in the cytosol and mitochondria to fold nascent and aggregated proteins. Hsp60 is the eukaryotic homolog of the E. coli GroEL protein, and forms a multimeric complex in the mitochondria with Hsp10 (Cpn10) to form a large central cavity in which ATP-dependent protein folding takes place. TRiC/CCT, a eukaryotic relative of Hsp60, is expressed in the cytosol and participates in the folding of actin and tubulin substrates, but lacks any association with an Hsp10-like co-factor.

  • For Immunofluorescence, Western Blot

Recognizes human, mouse, rat, bovine, dog, dolphin, Drosophila, fish, guinea pig, hamster, mosquito, monkey, pig, rabbit and snake HSP60. Detects a band of ~60kDa by Western blot.

Type: Primary
Antigen: HSP60
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR