Anti-CASP2 Rat Monoclonal Antibody [clone: 10C6]

Supplier: ProSci Inc.
36-178
PRSI36-178EA 568 GBP
PRSI36-178
Anti-CASP2 Rat Monoclonal Antibody [clone: 10C6]
Antibodies
Caspase-2 is a Class I caspase with a long prodomain necessary for nuclear localization. Upon activation of the apoptotic pathway, the procaspase is cleaved into smaller fragments. Caspase-2 is the nuclear apoptotic respondent to cellular genotoxic stress or mitotic catastrophe and is involved in the activation cascade of caspases responsible for apoptosis execution. Activation occurs upon recruitment to a complex containing a p53-induced death domain protein, PIDD. This suggests caspase-2 can be a nuclear initiator caspase with a requirement for caspase-9 and caspase-3 activation in downstream apoptotic events. In apoptotic pathways resulting from UV-induced DNA damage, processing of caspase-2 occurs downstream of mitochondrial dysfunction and of caspase-9 and caspase-3 activation, extending a possible role for caspase-2 as a parallel effector caspase.

Type: Primary
Antigen: CASP2
Clonality: Monoclonal
Clone: 10C6
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR