Anti-TNFRSF10D Mouse Monoclonal Antibody [clone: HS402]

Supplier: ProSci Inc.
36-135
PRSI36-135EA 667 GBP
PRSI36-135
Anti-TNFRSF10D Mouse Monoclonal Antibody [clone: HS402]
Antibodies
TRAIL-R4 is a receptor for the cytotoxic ligand TRAIL. Contains a truncated death domain and hence is not capable of inducing apoptosis. Protects cells against TRAIL mediated apoptosis by competing with TRAIL-R1 and R2 for binding to the ligand.

Type: Primary
Antigen: TNFRSF10D
Clonality: Monoclonal
Clone: HS402
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR