Human recombinant E-Selectin (from cells)
Supplier: ProSci Inc.
91-796
PRSI91-796EA 449 GBP
PRSI91-796
Human recombinant E-Selectin (from cells)
Proteins and Peptides
SELE is expressed on the surface of endothelial cells and mediates the interaction of leukocytes and platelets with endothelial cells during an inflammatory response. SELE functions as a cell-surface glycoprotein and has a role in immunoadhesion. In addition, SELE may also have a role in capillary morphogenesis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag