Anti-CRYGB Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
58-323
PRSI58-323EA 603 GBP
PRSI58-323
Anti-CRYGB Rabbit Polyclonal Antibody
Antibodies
For WB starting dilution is: 1:1000.

Type: Primary
Antigen: CRYGB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 68-97
Host: Rabbit
Isotype: Ig
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR