Anti-Transition Protein 1 Rabbit Polyclonal Antibody
Supplier: ProSci Inc.
30-898
PRSI30-898EA 642 GBP
PRSI30-898
Anti-Transition Protein 1 Rabbit Polyclonal Antibody
Antibodies
TNP1 is a spermatid-specific product of the haploid genome which replaces histone and is itself replaced in the mature sperm by the protamines.Transition protein-1 is a spermatid-specific product of the haploid genome which replaces histone and is itself replaced in the mature sperm by the protamines (see PRM1, MIM 182880; PRM2, MIM 182890) (Luerssen et al., 1990 [PubMed 2249851]).
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot andMore Product Information