Anti-KCNK13 Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
28-277
PRSI28-277EA 585 GBP
PRSI28-277
Anti-KCNK13 Rabbit Polyclonal Antibody
Antibodies
KCNK13 encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming domains. The product of KCNK13 is an open channel that can be stimulated by arachidonic acid.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: KCNK13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR