Anti-NKX3-2 Rabbit Polyclonal Antibody
Supplier: ProSci Inc.
27-134
PRSI27-134EA 642 GBP
PRSI27-134
Anti-NKX3-2 Rabbit Polyclonal Antibody
Antibodies
NKX3-2 is a member of the NK family of homeobox-containing proteins. It may play a role in skeletal development.This gene encodes a member of the NK family of homeobox-containing proteins. The encoded protein may play a role in skeletal development. Sequence Note: This RefSeq record was created from transcript and genomic sequence data because no single transcript was available for the full length of the gene. The extent of this transcript is supported by transcript alignments.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2%More Product Information