Anti-RNF115 Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
25-828
PRSI25-828EA 642 GBP
PRSI25-828
Anti-RNF115 Rabbit Polyclonal Antibody
Antibodies
ZNF364 contains 1 RING-type zinc finger. The exact function of ZNF364 remains unknown.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RNF115
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR