Masterflex® Tank Dispensing Spray Guns, Avantor®
Supplier: Avantor Fluid Handling
98515-01 98515-03 98515-05 98515-07
MFLX98515-01EA 214 GBP
MFLX98515-01 MFLX98515-03 MFLX98515-05 MFLX98515-07
Masterflex® Tank Dispensing Spray Guns, Avantor®
Spray Guns
No splashing.
  • Recirculating gun has a contamination free fluid path to eliminate bacterial growth
  • Highly resistant to corrosive substances and fumes
  • Spray guns have a 1 GPM flow rate; max operating pressure of 75 psi
These Masterflex® Tank Dispensing Spray Guns feature a special tip to eliminate splashing. Choose from PTFE or stainless steel construction for a wide range of applications.