Masterflex® Spray Gun, PTFE for DI Water, 1/2" NPT(F); 1/Ea
Supplier: Avantor Fluid Handling
VWR Item#MFLX98515-03
Your Organization's Price:
Retrieving
No splashing.
  • Recirculating gun has a contamination free fluid path to eliminate bacterial growth
  • Highly resistant to corrosive substances and fumes
  • Spray guns have a 1 GPM flow rate; max operating pressure of 75 psi
These Masterflex® Tank Dispensing Spray Guns feature a special tip to eliminate splashing. Choose from PTFE or stainless steel construction for a wide range of applications.