Anti-IL3RA Mouse Monoclonal Antibody (Biotin) [clone: 107D2.08]

Supplier: ENZO LIFE SCIENCES
ENZABS2310100
ENZOENZABS2310100EA 561 GBP
ENZOENZABS2310100
Anti-IL3RA Mouse Monoclonal Antibody (Biotin) [clone: 107D2.08]
Antibodies
Recommended Applications: IHC (PS), IP

Species reactivity: Human, Dog

Type: Primary
Antigen: IL3RA
Clonality: Monoclonal
Clone: 107D2.08
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR