Nuclear-ID™ red cell cycle kit, GFP-CERTIFIED®, for flow cytometry
Supplier: ENZO LIFE SCIENCES
ENZ-51008-100 ENZ51008100
ALEXENZ-51008-100EA 541 GBP
ALEXENZ-51008-100 ENZOENZ51008100
Nuclear-ID™ red cell cycle kit, GFP-CERTIFIED®, for flow cytometry
Assays Cellular Assays
GFP-Certified™ Nuclear-ID™ Red Cell Cycle Analysis Kit provides a convenient approach for studying the induction and inhibition of cell cycle progression by flow cytometry.
The kit is suitable for determining the percentage of cells in a given sample that are in G0/G1, S and G2/M phases, as well as to quantify cells in the sub-G1 phase, and DNA studies in live, permeabilised and fixed cells for normal cell lines and cell lines exhibiting multiple ploidy levels. A control cell cycle perturbation agent, Nocodazole, is provided for monitoring changes in cell cycle dynamics. Potential applications for live-cell studies are in the determination of cellular DNA content and cell cycle distribution for the detection of variations in growth patterns, for monitoring apoptosis, and for evaluating tumor cell behaviour and suppressor gene mechanisms.
Ordering information: Kit contains: Nuclear-ID™ Red Cell Cycle Detection Reagent, 200 µl; Nocodazole Control, 10 µl; 10X Assay Buffer, 15 ml.
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable
Application Fluorescence Microscopy, Fluorescent Detection
Size 1 kit