Blur 8 BioProbe®
Supplier: ENZO LIFE SCIENCES
ENZ-40849 ENZ40849
ALEXENZ-40849EA 455 GBP
ALEXENZ-40849 ENZOENZ40849
Blur 8 BioProbe®
Nucleic Acid Reagents Primers and Probes
The Blur 8 BioProbe® labeled probe is prepared by nick translation of a clone of the human alu repeat sequence. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis).