Shp 1 and 2 substrate
Supplier: ENZO LIFE SCIENCES
BML-P157-0001 BMLP1570001
ALEXBML-P157-0001EA 189 GBP
ALEXBML-P157-0001 ENZOBMLP1570001
Shp 1 and 2 substrate
Proteins and Peptides
This peptide sequence, EFpYAEVGRSPPDPAK, was selected by phage display analysis for optimal Shp-1 and Shp-2 phosphatase activities.