Cathepsin S drug discovery kit
Supplier: ENZO LIFE SCIENCES
BML-AK431-0001 BMLAK4310001
ALEXBML-AK431-0001EA 615 GBP
ALEXBML-AK431-0001 ENZOBMLAK4310001
Cathepsin S drug discovery kit
Assays Enzyme Assays
A QuantiZyme™ Assay System. The Cathepsin S Drug Discovery Kit is a complete assay system designed to screen inhibitors of cathepsin S. Using a convenient microplate format, cathepsin S activity is monitored fluorimetrically using the substrate provided in the kit.
  • Complete system designed to screen modulators of cathepsin S activity
  • The assays are performed in a convenient 96-well microplate format
Delivery information: Kit contains Cathepsin S enzyme (human) (recombinant), AMC substrate, AMC calibration standard, inhibitor, assay buffer and ½-volume white microplate.