Live-dead cell staining kit
Supplier: ENZO LIFE SCIENCES
ALX850249KI01
ENZOALX850249KI01EA 441 GBP
ENZOALX850249KI01
Live-dead cell staining kit
Assays Cellular Assays
Kit utilises Live-dye™, a cell permeable green fluorescent dye (Ex(max): 488 nm; Em(max): 518 nm), to stain live cells. Dead cells can be easily stained by propidium iodide (PI), a red fluorescent dye (Ex(max): 488 nm; Em(max): 615 nm), which in viable cells is actively pumped out of the cytoplasm.
  • Ready-to-use
  • Live-dead cell stain for 100 tests
Stained cells can be visualised by fluorescence microscopy using a band-pass filter (detects FITC and rhodamine). Cyto-dye has also been used to discriminate between apoptotic and non-apoptotic cells.