omega-Agatoxin IVA
Supplier: ENZO LIFE SCIENCES
ALX630001C100
ENZOALX630001C100EA 727 GBP
ENZOALX630001C100
omega-Agatoxin IVA
Proteins and Peptides
Ca²⁺ channel blocker
Selective P-type Ca²⁺ channel blocker. Fast acting and potent neurotoxin.