Ruthenium(III) chloride oxide 90 Metals basis

Supplier: ENZO LIFE SCIENCES

Synonyms: Ruthenium oxychloride, Tetraamminechlorohydroxoruthenium(III) chloride, Ruthenium Red

ALX550295G001
ENZOALX550295G001EA 167 GBP
ENZOALX550295G001
Ruthenium(III) chloride oxide 90 Metals basis
Ruthenium(III) chloride oxide

Blocks TRPV1. Capsaicin and calcium antagonist. Inhibitor of Ca²⁺/Mg2⁺-ATPase.


  • Capsaicin and Ca2⁺ antagonist


Dark red to purple crystalline solid.


Soluble in water (10mg/ml) or ammonia.

Formula: H₄₂Cl₆N₁₄O₂Ru₃
MW: 786.354 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011479
CAS Number: 11103-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR