Tamoxifen citrate 99

Supplier: ENZO LIFE SCIENCES

Synonyms: (Z)-1-(4-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene Citrate

ALX550095G001
ENZOALX550095G001EA 181 GBP
ENZOALX550095G001
Tamoxifen citrate 99
Tamoxifen citrate

PKC inhibitor


Antiestrogen with anti-cancer activity. Selective protein kinase C (PKC) inhibitor and induces PKC epsilon translocation. Induces apoptosis in estrogen receptor-negative human cancer cell lines.

Formula: C₂₆H₂₉NO
MW: 563.65 g/mol
MDL Number: MFCD00058321
CAS Number: 54965-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR