Nadifloxacin ≥98%

Supplier: ENZO LIFE SCIENCES

Synonyms: 9-Fluoro-8-(4-hydroxypiperidin-1-yl)-5-methyl-1-oxo-1,5,6,7-tetrahydropyrido[3,2,1-ij]quinoline-2-carboxylic acid, 9-Fluoro-8-(4-hydroxy-1-piperidinyl)-5-methyl-1-oxo-1,5,6,7-tetrahydropyrido[3,2,1-ij]quinoline-2-carboxylic Acid

ALX380294G001
ENZOALX380294G001EA 433 GBP
ENZOALX380294G001
Nadifloxacin ≥98%
Nadifloxacin

Fluoroquinolone with broad spectrum activity against Gram-negative and Gram-positive bacteria. Inhibits the generation of O2- and OH radicals by neutrophils. Inhibits the production of inflammatory cytokines.


  • White to yellow crystalline solid


Soluble in DMSO


Packaging: Shipping temperature: Ambient

Formula: C₁₉H₂₁FN₂O₄
MW: 360.39 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00865081
CAS Number: 124858-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR