Anti-GSK3A Mouse Monoclonal Antibody

Supplier: ENZO LIFE SCIENCES
ADI905759100
ENZOADI905759100EA 526 GBP
ENZOADI905759100
Anti-GSK3A Mouse Monoclonal Antibody
Antibodies
Glycogen Synthase Kinase 3 (GSK3) exists in two isoforms, alpha and beta (GSK3α, and GSK3β). GSK3 is an active kinase that becomes phosphorylated by PKB/akt upon growth factor stimulation. Phosphorylation of GSK3α at serine 21 or GSK3β at serine 9, respectively, leads to inhibition of kinase activity of the GSK3.

  • For ELISA, Flow Cytometry, Western Blot

Recognizes human, mouse, rat and dog GSK-3alpha.

Reconstitute with 1ml water (15 minutes at room temperature).

Type: Primary
Antigen: GSK-3ALPHA
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR