Anti-IL8 Mouse Monoclonal Antibody [clone: NYR-HIL8]

Supplier: ENZO LIFE SCIENCES
ADI905110500
ENZOADI905110500EA 409 GBP
ENZOADI905110500
Anti-IL8 Mouse Monoclonal Antibody [clone: NYR-HIL8]
Antibodies
Interleukin-8 (IL-8) is a cytokine produced by monocytes, macrophages, neutrophils, lymphocytes, dermal fibroblasts, keratinocytes, vascular endothelial cells, melanocytes, hepatocytes and various tumor cell lines. It is also a member of the CXC family of chemokines, displaying chemotactic activity for neutrophils and T cells.

Type: Primary
Antigen: IL8
Clonality: Monoclonal
Clone: NYR-HIL8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol IL8
Environmentally Preferable
Antigen name Interleukin 8
Antigen synonyms MDNCF|Interleukin-8|T-cell chemotactic factor|LYNAP|NAP1|NAP-1|GCP1|Monocyte-derived neutrophil chemotactic factor|MONAP|LECT|NAF|GCP-1|CXCL8|LUCT|Neutrophil-activating protein 1
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone NYR-HIL8
Reactivity Human
Host Mouse
Isotype IgG1
Western blot Yes
ELISA Yes
ImmunoPrecipitation Yes
Immunogen Recombinant human Interleukin-8.
Form Lyophilized
Purification purified by ion exchange chromatography
Storage buffer PBS
Storage temperature 4 °C
Shipping temperature Blue ice not frozen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR