GRO/CINC-1 (Rat), ELISA kit

Supplier: ENZO LIFE SCIENCES

ADI-900-074 ADI900074
ASSAADI-900-074EA 845 GBP
ASSAADI-900-074 ENZOADI900074
GRO/CINC-1 (Rat), ELISA kit
Assays ELISAs

The GRO/CINC-1 (Rat), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 2 hours. Absorbance is read at 450 nm. Assay up to 39 samples in duplicate per kit.


  • Fast and fully quantitative results
  • Sensitive
  • Higher throughput


For the quantitative determination of rat GRO/CINC-1 in culture supernatants, plasma, and serum.


Delivery information: Supplied with microtiter plate, standard, antibody, conjugate, assay buffer 28, TMB substrate, stop solution 2, wash buffer concentrate.

New Product

Order Now

SPECIFICATIONS

Target protein GRO/CINC-1
Environmentally Preferable
Primary antibody reactivity Rat
Host
Sensitivity 1,99 pg/ml (range 4,7 - 300 pg/ml)
Storage conditions 4 °C (long term)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR