Film, Kodak BioMax® XAR
Supplier: CARESTREAM HEALTH
1651496 1651512 1651579 1651678 1660760
730-3216EA 0 GBP
730-3216 730-3217 730-3218 730-3219 730-3221
Film, Kodak BioMax® XAR
Autoradiography Films Films
BioMax® XAR Film is an autoradiography film for use with all commonly used isotopes and chemiluminescence labels. The film is coated with emulsion on both sides of a clear base, and is suitable for both direct autoradiography and exposures made with amplification screens, including BioMax® amplification screens.
  • Suitable for both manual and automated processing
Darkroom handling: BioMax® XAR film should be handled under safelight conditions using a safelight lamp equipped with a Safelight Filter GBX-2 and 15 W frosted bulb (other safelight filters mayMore Product Information