Film, Amersham™ Hyperfilm™ ECL™
Supplier: Cytiva
28-9068-36 28-9068-37 28-9068-35 28-9068-39 28-9068-38
28-9068-36EA 324 GBP
28-9068-36 28-9068-37 28-9068-35 28-9068-39 28-9068-38
Film, Amersham™ Hyperfilm™ ECL™
Autoradiography Films Films
Amersham™ Hyperfilm™ ECL™ is constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti-scratch layer. The anti-scratch coating reduces the risk of visible artefacts (e.g., plastic wrap marks) and the clear colourless base results in easy reading of images without compromising on sensitivity.
  • High sensitivity for low concentration of nucleic acid and protein targets
  • Optimised for use in all blue and green light chemiluminescence systems
  • Clear background for excellent contrast and improved band resolution
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable