Vacuum gauge set DCP 3000 + VSP 3000
Supplier: VACUUBRAND
20683190 20636155
181-0174EA 1320 GBP
181-0174 VACU20636155
Vacuum gauge set DCP 3000 + VSP 3000
Pumps Vacuum Gauges
The DCP 3000 is an outstanding vacuum gauge with large illuminated display. The brand new VSP 3000 vacuum sensor is based on thermal conductivity (Pirani) and offers a wide measurement range from atmospheric pressure down to 10 to 3 mbar. The wetted parts are made of chemically resistant plastics (PBT, PUR) and alumina ceramics. It features a much better chemical resistance and ruggedness than conventional Pirani sensors with their fragile metallic spiral-wound filament. Simultaneous connection of up to eight vacuum sensors allows flexible measuring on the spot. The DCP 3000 communicatesMore Product Information